ENA112933061 Enamine

{[(6-methylimidazo[1,2-a]pyridin-2-yl)methyl]thio}acetic acid

  • MDL number MFCD09040431
  • Cas Number : 929975-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26848

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26848
Availability details for
Quantity
  • Availability
  • Add to Cart