ENA112933062 Enamine

ethyl 5-amino-1-[(tert-butoxycarbonyl)amino]-4-cyano-2-methyl-1H-pyrrole-3-carboxylate

  • MDL number MFCD09040433
  • Cas Number : 129378-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26859
Availability details for
Quantity
  • Availability
  • Add to Cart