ENA112933066 Enamine

(3S)-6,6-dimethyl-5-oxothiomorpholine-3-carboxylic acid

  • MDL number MFCD09040462
  • Cas Number : 79438-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26934
Availability details for
Quantity
  • Availability
  • Add to Cart