ENA112933073 Enamine

2-[3-(furan-2-yl)-5-sulfanyl-4H-1,2,4-triazol-4-yl]acetamide

  • MDL number MFCD09040469
  • Cas Number : 929975-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26946
Availability details for
Quantity
  • Availability
  • Add to Cart