ENA112933080 Enamine

tert-butyl N-[3-(thiophen-2-yl)prop-2-yn-1-yl]carbamate

  • MDL number MFCD09040478
  • Cas Number : 889451-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26972
Availability details for
Quantity
  • Availability
  • Add to Cart