ENA112933083 Enamine

1-(2-methoxyethyl)-4,5-dimethyl-1H-imidazole-2-thiol

  • MDL number MFCD09040482
  • Cas Number : 929975-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27020

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27020
Availability details for
Quantity
  • Availability
  • Add to Cart