ENA112933090 Enamine

4-methyl-5-(pyridin-2-yl)-2,3-dihydro-1,3-thiazol-2-imine

  • MDL number MFCD23146398
  • Cas Number : 94284-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27039
Availability details for
Quantity
  • Availability
  • Add to Cart