ENA112933093 Enamine

1-(1-bicyclo[2.2.1]hept-2-ylethyl)-4,5-dimethyl-1H-imidazole-2-thiol

  • MDL number MFCD13806374
  • Cas Number : 1005044-09-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27057
Availability details for
Quantity
  • Availability
  • Add to Cart