ENA112933095 Enamine

2-bromo-6-tert-butylimidazo[2,1-b][1,3,4]thiadiazole

  • MDL number MFCD09040495
  • Cas Number : 929975-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27059

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27059
Availability details for
Quantity
  • Availability
  • Add to Cart