ENA112933099 Enamine

1-[4-(5-bromothiophen-2-yl)phenyl]ethan-1-one

  • MDL number MFCD09040499
  • Cas Number : 929972-12-3
  • Molecular Weight : 281.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27073
Availability details for
Quantity
  • Availability
  • Add to Cart