ENA112933100 Enamine

ethyl 3-cyano-2-hydroxy-2-(1-methyl-1H-imidazol-2-yl)propanoate

  • MDL number MFCD09040500
  • Cas Number : 929972-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27074
Availability details for
Quantity
  • Availability
  • Add to Cart