ENA112933116 Enamine

1-ethyl-1H-imidazole-2-thiol

  • MDL number MFCD08691180
  • Cas Number : 10583-83-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26018

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26018
Availability details for
Quantity
  • Availability
  • Add to Cart