ENA112933117 Enamine

1-cyclopropyl-1H-imidazole-2-thiol

  • MDL number MFCD08691182
  • Cas Number : 923156-13-2
  • Molecular Weight : 140.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26020
Availability details for
Quantity
  • Availability
  • Add to Cart