ENA112933120 Enamine

6-(1H-1,2,4-triazol-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD01936059
  • Cas Number : 400075-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26076
Availability details for
Quantity
  • Availability
  • Add to Cart