ENA112933123 Enamine

6-chloropyridazine-3-thiol

  • MDL number MFCD08691217
  • Cas Number : 3916-78-7
  • Molecular Weight : 146.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 160-162
Iupac Name 6-chloro-3(2H)-pyridazinethione
Inchi Code 1S/C4H3ClN2S/c5-3-1-2-4(8)7-6-3/h1-2H,(H,7,8)
Inchi Key ABQHEWLZYZQXHY-UHFFFAOYSA-N

Description

Melting Point 160-162
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26124

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-26124
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart