ENA112933133 Enamine

3-(1H-1,2,4-triazol-1-yl)propanoic acid

  • MDL number MFCD02932735
  • Cas Number : 76686-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178...180
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26147

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26147
Availability details for
Quantity
  • Availability
  • Add to Cart