ENA112933138 Enamine

6-(4-methylpiperazin-1-yl)nicotinonitrile

  • MDL number MFCD01692399
  • Cas Number : 54864-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26156

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26156
Availability details for
Quantity
  • Availability
  • Add to Cart