ENA112933150 Enamine

5-ethoxy-2-(4-fluorophenyl)-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD08691230
  • Cas Number : 923819-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26172
Availability details for
Quantity
  • Availability
  • Add to Cart