ENA112933160 Enamine

N-(4-{[4-(hydroxyimino)piperidin-1-yl]sulfonyl}phenyl)acetamide

  • MDL number MFCD08691233
  • Cas Number : 923108-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200...202
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26183

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26183
Availability details for
Quantity
  • Availability
  • Add to Cart