ENA112933162 Enamine

4-(morpholin-4-yl)-3-nitro-2H-chromen-2-one

  • MDL number MFCD00420263
  • Cas Number : 50527-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26185
Availability details for
Quantity
  • Availability
  • Add to Cart