ENA112933174 Enamine

1-(4-chlorophenyl)propan-1-amine

  • MDL number MFCD08691240
  • Cas Number : 74788-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26199

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26199
Availability details for
Quantity
  • Availability
  • Add to Cart