ENA112933186 Enamine

4-amino-3-nitro-2H-chromen-2-one

  • MDL number MFCD00126356
  • Cas Number : 38464-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26223

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26223
Availability details for
Quantity
  • Availability
  • Add to Cart