ENA112933188 Enamine

2-cyclopropyl-5-nitro-1H-1,3-benzodiazole

  • MDL number MFCD08691249
  • Cas Number : 51758-97-5
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26225
Availability details for
Quantity
  • Availability
  • Add to Cart