ENA112933189 Enamine

2-[4-(hydroxyimino)piperidin-1-yl]-N-phenylacetamide

  • MDL number MFCD08691250
  • Cas Number : 923216-32-4
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26226
Availability details for
Quantity
  • Availability
  • Add to Cart