ENA112933190 Enamine

2-[(4-chloro-1,3-thiazol-2-yl)sulfanyl]benzoic acid

  • MDL number MFCD08691251
  • Cas Number : 923797-62-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26227
Availability details for
Quantity
  • Availability
  • Add to Cart