ENA112933192 Enamine

1-[(furan-2-yl)methyl]-5-phenyl-1H-imidazole-2-thiol

  • MDL number MFCD08691252
  • Cas Number : 923862-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26230

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26230
Availability details for
Quantity
  • Availability
  • Add to Cart