ENA112933197 Enamine

2-(furan-2-amido)-3-methylbenzoic acid

  • MDL number MFCD08691256
  • Cas Number : 923784-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-26240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26240
Availability details for
Quantity
  • Availability
  • Add to Cart