ENA112933203 Enamine

1-methyl-1H,2H,3H,4H-pyrazino[2,3-b]quinoxaline

  • MDL number MFCD08689813
  • Cas Number : 923689-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26248
Availability details for
Quantity
  • Availability
  • Add to Cart