ENA112933207 Enamine

(1E)-2-(1H-benzimidazol-2-yl)-1-phenylethanone oxime

  • MDL number MFCD01878769
  • Cas Number : 451515-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26252
Availability details for
Quantity
  • Availability
  • Add to Cart