ENA112933209 Enamine

4-[5-(1-chloroethyl)-1,2,4-oxadiazol-3-yl]pyridine

  • MDL number MFCD08691260
  • Cas Number : 923232-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26254
Availability details for
Quantity
  • Availability
  • Add to Cart