ENA112933212 Enamine

1-(2-furoyl)piperidin-4-one oxime

  • MDL number MFCD08691262
  • Cas Number : 923232-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26257

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26257
Availability details for
Quantity
  • Availability
  • Add to Cart