ENA112933220 Enamine

3-(carbamoylamino)-3-(4-methylphenyl)propanoic acid

  • MDL number MFCD08691263
  • Cas Number : 923215-91-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-211
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26265
Availability details for
Quantity
  • Availability
  • Add to Cart