ENA112933226 Enamine

1-(4-chlorophenyl)-2-methylpropan-1-amine

  • MDL number MFCD08691269
  • Cas Number : 78469-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26273
Availability details for
Quantity
  • Availability
  • Add to Cart