ENA112933233 Enamine

2-(phenoxymethyl)benzonitrile

  • MDL number MFCD08070461
  • Cas Number : 168971-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26280

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26280
Availability details for
Quantity
  • Availability
  • Add to Cart