ENA112933248 Enamine

2-phenyl-2-[(thiophen-2-yl)formamido]acetic acid

  • MDL number MFCD04971917
  • Cas Number : 1008670-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-26305

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26305
Availability details for
Quantity
  • Availability
  • Add to Cart