ENA112933252 Enamine

ethyl 5-(chlorosulfonyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD08691284
  • Cas Number : 115703-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26314

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26314
Availability details for
Quantity
  • Availability
  • Add to Cart