ENA112933266 Enamine

1-{[3-(4-chlorophenyl)-1,2,4-oxadiazol-5-yl]methyl}piperazine

  • MDL number MFCD08691296
  • Cas Number : 923721-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236...238
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26331

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26331
Availability details for
Quantity
  • Availability
  • Add to Cart