ENA112933272 Enamine

4-(5-phenyl-1,3,4-oxadiazol-2-yl)benzoic acid

  • MDL number MFCD00491629
  • Cas Number : 85292-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 302-304
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26338
Availability details for
Quantity
  • Availability
  • Add to Cart