ENA112933279 Enamine

2-[(4-phenyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetic acid

  • MDL number MFCD00227618
  • Cas Number : 3682-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185-186
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26349
Availability details for
Quantity
  • Availability
  • Add to Cart