ENA112933283 Enamine

2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-sulfonyl chloride

  • MDL number MFCD05863680
  • Cas Number : 952-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 290-292
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26356
Availability details for
Quantity
  • Availability
  • Add to Cart