ENA112933286 Enamine

2-(1-methyl-1H-benzimidazol-2-yl)-1-phenylethanone

  • MDL number MFCD08691304
  • Cas Number : 58112-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26362

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26362
Availability details for
Quantity
  • Availability
  • Add to Cart