ENA112933288 Enamine

N-{3-[1-(methylamino)ethyl]phenyl}benzamide

  • MDL number MFCD08691306
  • Cas Number : 923783-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26367
Availability details for
Quantity
  • Availability
  • Add to Cart