ENA112933291 Enamine

sodium 3-amino-4,6-dimethylthieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD08691308
  • Cas Number : 144254-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 291...293
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26372
Availability details for
Quantity
  • Availability
  • Add to Cart