ENA112933303 Enamine

2-[4-(4-methylphenyl)-2-oxo-2,3-dihydro-1,3-thiazol-3-yl]acetic acid

  • MDL number MFCD08691317
  • Cas Number : 923105-85-5
  • Molecular Weight : 249.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-129
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26388
Availability details for
Quantity
  • Availability
  • Add to Cart