ENA112933312 Enamine

3-[5-(2,4-difluorophenyl)-1,3-oxazol-2-yl]propanoic acid

  • MDL number MFCD08691323
  • Cas Number : 923825-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26401
Availability details for
Quantity
  • Availability
  • Add to Cart