ENA112933317 Enamine

2-[2-(2-ethoxyphenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06857879
  • Cas Number : 923751-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26407
Availability details for
Quantity
  • Availability
  • Add to Cart