ENA112933318 Enamine

5-benzyl-4-[(furan-2-yl)methyl]-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691328
  • Cas Number : 537017-46-2
  • Molecular Weight : 271.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26408
Availability details for
Quantity
  • Availability
  • Add to Cart