ENA112933319 Enamine

5-(2-cyclopentylethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691329
  • Cas Number : 911208-58-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26409
Availability details for
Quantity
  • Availability
  • Add to Cart