ENA112933321 Enamine

3-[5-(2-fluorophenyl)-1,3-oxazol-2-yl]propanoic acid

  • MDL number MFCD08691331
  • Cas Number : 923108-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26411
Availability details for
Quantity
  • Availability
  • Add to Cart