ENA112933326 Enamine

4-(hydroxyimino)-N-phenylpiperidine-1-carboxamide

  • MDL number MFCD08691335
  • Cas Number : 923163-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26419

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26419
Availability details for
Quantity
  • Availability
  • Add to Cart